3-[2-(2-Ethoxycarbonyl-ethoxy)-ethoxy]-propionic acid 97%

Lieferant: Apollo Scientific

OR964496-1G OR964496-250MG
APOSOR964496-1GEA 0 EUR
APOSOR964496-1G APOSOR964496-250MG
3-[2-(2-Ethoxycarbonyl-ethoxy)-ethoxy]-propionic acid 97%
3-[2-(2-Ethoxycarbonyl-ethoxy)-ethoxy]-propionic acid
Formel: C₁₀H₁₈O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD04115349
CAS-Nummer: 886362-90-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR