5-Methyl-1H-pyrrolo[2,3-b]pyridine 98%

Lieferant: Apollo Scientific

OR964572-1G
APOSOR964572-1GEA 59.6 EUR
APOSOR964572-1G
5-Methyl-1H-pyrrolo[2,3-b]pyridine 98%
5-Methyl-1H-pyrrolo[2,3-b]pyridine
Formel: C8H8N2
Molecular Weight: 132.1625 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659674
CAS-Nummer: 824-52-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR