(3-Iodo-2,5,6-trimethoxypyridin-4-yl)boronic acid

Lieferant: Apollo Scientific

OR964620-1G
APOSOR964620-1GEA 1740 EUR
APOSOR964620-1G
(3-Iodo-2,5,6-trimethoxypyridin-4-yl)boronic acid
(3-Iodo-2,5,6-trimethoxypyridin-4-yl)boronic acid
Formel: C₈H₁₁BINO₅
Molecular Weight: 338.892 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22887387
CAS-Nummer: 2096338-48-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR