4-Hydroxybenzooxazole-2(3H)-thione 95%

Lieferant: Apollo Scientific

OR964659-1G
APOSOR964659-1GEA 632 EUR
APOSOR964659-1G
4-Hydroxybenzooxazole-2(3H)-thione 95%
4-Hydroxybenzooxazole-2(3H)-thione
Formel: C₇H₅NO₂S
Molecular Weight: 167,185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16495908
CAS-Nummer: 73713-92-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR