4-Ethoxy-2-hydroxy-benzaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 4-Ethoxy-2-hydroxybenzaldehyde

OR964660-1G
APOSOR964660-1GEA 102 EUR
APOSOR964660-1G
4-Ethoxy-2-hydroxy-benzaldehyde 95%
4-Ethoxy-2-hydroxy-benzaldehyde
Formel: C₉H₁₀O₃
Molecular Weight: 166,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03032734
CAS-Nummer: 43057-77-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR