3-(Aminomethyl)-N,N-dimethylbenzamide hydrochloride 97%

Lieferant: Apollo Scientific

Synonyme: 3-(Aminomethyl)-N,N-dimethyl-benzamide

OR964687-1G OR964687-250MG
APOSOR964687-1GEA 464 EUR
APOSOR964687-1G APOSOR964687-250MG
3-(Aminomethyl)-N,N-dimethylbenzamide hydrochloride 97%
3-(Aminomethyl)-N,N-dimethylbenzamide hydrochloride
Formel: C₁₀H₁₅ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09836256
CAS-Nummer: 863548-47-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR