3-(Aminomethyl)-N,N-dimethylbenzamide hydrochloride 97%
Lieferant: Apollo Scientific
Synonyme:
3-(Aminomethyl)-N,N-dimethyl-benzamide
OR964687-1G
OR964687-250MG
APOSOR964687-1GEA
464
EUR
APOSOR964687-1G
APOSOR964687-250MG
3-(Aminomethyl)-N,N-dimethylbenzamide hydrochloride 97%
3-(Aminomethyl)-N,N-dimethylbenzamide hydrochloride
Formel:
C₁₀H₁₅ClN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD09836256 CAS-Nummer: 863548-47-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...