1,2-Phenylendiessigsäure 95%
Lieferant: Apollo Scientific
OR964730-25G
APOSOR964730-25GEA
56.7
EUR
APOSOR964730-25G
1,2-Phenylendiessigsäure 95%
1,2-Phenylendiessigsäure
Formel:
C₁₀H₁₀O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004329 CAS-Nummer: 7500-53-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...