2-Amino-4-(chloromethyl)pyridine hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-4-(chloromethyl)pyridine Hydrochloride

OR964731-1G
APOSOR964731-1GEA 134 EUR
APOSOR964731-1G
2-Amino-4-(chloromethyl)pyridine hydrochloride 95%
2-Amino-4-(chloromethyl)pyridine hydrochloride
Formel: C₆H₇ClN₂·ClH
Molecular Weight: 179.047 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12198864
CAS-Nummer: 1186663-31-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR