5-Methoxy-7-azaindole 97%

Lieferant: Apollo Scientific

Synonyme: 5-Methoxy-1H-pyrrolo[2,3-b]pyridine

OR964856-5G
APOSOR964856-5GEA 87 EUR
APOSOR964856-5G
5-Methoxy-7-azaindole 97%
5-Methoxy-7-azaindole
Formel: C8H8N2O
Lagertemperatur: Raumtemperatur
MDL: MFCD06659675
CAS-Nummer: 183208-36-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR