Methyl 2-(1-methylpiperidin-4-yl)acetate 98%

Lieferant: Apollo Scientific

OR964903-1G
APOSOR964903-1GEA 342 EUR
APOSOR964903-1G
Methyl 2-(1-methylpiperidin-4-yl)acetate 98%
Methyl 2-(1-methylpiperidin-4-yl)acetate
Formel: C₉H₁₇NO₂
Molecular Weight: 171,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12756507
CAS-Nummer: 95533-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR