Ethyl-3-amino-5-chlorbenzoat 95%
Lieferant: Apollo Scientific
OR964922-1G
APOSOR964922-1GEA
348
EUR
APOSOR964922-1G
Ethyl-3-amino-5-chlorbenzoat 95%
Ethyl-3-amino-5-chlorbenzoat
Formel:
C₉H₁₀ClNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD17276491 CAS-Nummer: 1261269-01-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...