4-Isopropyl-1,3-thiazol-2-amine hydrobromide 98%

Lieferant: Apollo Scientific

Synonyme: 4-Isopropyl-1,3-thiazol-2-amine

OR965018-5G
APOSOR965018-5GEA 89.9 EUR
APOSOR965018-5G
4-Isopropyl-1,3-thiazol-2-amine hydrobromide 98%
4-Isopropyl-1,3-thiazol-2-amine hydrobromide
Formel: C₆H₁₁BrN₂S
Molecular Weight: 223,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21605754
CAS-Nummer: 79932-20-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR