Ethyl 3-Oxo-3-(pyrazin-2-yl)propionate 98%

Lieferant: Apollo Scientific

OR965038-5G
APOSOR965038-5GEA 108 EUR
APOSOR965038-5G
Ethyl 3-Oxo-3-(pyrazin-2-yl)propionate 98%
Ethyl 3-Oxo-3-(pyrazin-2-yl)propionate
Formel: C₉H₁₀N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04114374
CAS-Nummer: 62124-77-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR