2-Hydroxyethanesulphonic acid 80% in Wasser 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Isethionic acid

OR965087-100G OR965087-500G
APOSOR965087-100GEA 102 EUR
APOSOR965087-100G APOSOR965087-500G
2-Hydroxyethanesulphonic acid 80% in Wasser 97%
2-Hydroxyethanesulphonic acid
Formel: C₂H₆O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD00242599
CAS-Nummer: 107-36-8
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR