2-Hydroxyethanesulphonic acid 80% in Wasser 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Isethionic acid
OR965087-100G
OR965087-500G
APOSOR965087-100GEA
102
EUR
APOSOR965087-100G
APOSOR965087-500G
2-Hydroxyethanesulphonic acid 80% in Wasser 97%
2-Hydroxyethanesulphonic acid
Formel:
C₂H₆O₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00242599 CAS-Nummer: 107-36-8 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...