Azomethin H

Lieferant: Apollo Scientific

Synonyme: 8-Hydroxy-1-[[(2-hydroxyphenyl)methylen]amino]naphthalin-3,6-disulfonsäure, Azomethine-H, 8-Hydroxy-1-(salicylidenamino)naphthalen-3,6-disulfonsäure, 4-Hydroxy-5-[[(2-hydroxyphenyl)methylen]amino]naphthalin-2,7-disulfonsäure, 4-Hydroxy-5-(salicylidenamino)naphthalin-2,7-disulfonsäure

OR9650T-5G
APOSOR9650T-5GEA 105 EUR
APOSOR9650T-5G
Azomethin H
Azomethin H

A highly sensitive reagent for testing boron.

Formel: C₁₇H₁₃NO₈S₂
Molecular Weight: 423,42 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 32266-60-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR