2-Oxo-2,5,6,7-tetrahydro-1H-[1]pyrindine-3-carboxylic acid 95%
Lieferant: Apollo Scientific
Synonyme:
2-Hydroxy-6,7-dihydro-5h-cyclopenta[b]pyridine-3-carboxylic acid
OR965109-1G
APOSOR965109-1GEA
74.1
EUR
APOSOR965109-1G
2-Oxo-2,5,6,7-tetrahydro-1H-[1]pyrindine-3-carboxylic acid 95%
2-Oxo-2,5,6,7-tetrahydro-1H-[1]pyrindine-3-carboxylic acid
Formel:
C₉H₉NO₃ Molecular Weight: 179,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07801152 CAS-Nummer: 115122-63-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...