3-Methoxy-2-cyclopenten-1-one 95%

Lieferant: Apollo Scientific

Synonyme: 3-Methoxycyclopent-2-enone

OR965147-1G
APOSOR965147-1GEA 114 EUR
APOSOR965147-1G
3-Methoxy-2-cyclopenten-1-one 95%
3-Methoxy-2-cyclopenten-1-one
Formel: C₆H₈O₂
Molecular Weight: 112,127 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00192267
CAS-Nummer: 4683-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR