3-Methylbenzo[1,2,4]triazine 97%
Lieferant: Apollo Scientific
OR965203-1G
APOSOR965203-1GEA
560
EUR
APOSOR965203-1G
3-Methylbenzo[1,2,4]triazine 97%
3-Methylbenzo[1,2,4]triazine
Formel:
C₈H₇N₃ Molecular Weight: 145.161 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00100923 CAS-Nummer: 6299-94-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...