3-Oxo-3,4-dihydro-2H-benzo[1,4]oxazine-6-carbaldehyde 95%

Lieferant: Apollo Scientific

OR965275-1G
APOSOR965275-1GEA 400 EUR
APOSOR965275-1G
3-Oxo-3,4-dihydro-2H-benzo[1,4]oxazine-6-carbaldehyde 95%
3-Oxo-3,4-dihydro-2H-benzo[1,4]oxazine-6-carbaldehyde
Formel: C₉H₇NO₃
Molecular Weight: 177,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07369979
CAS-Nummer: 200195-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR