Ethyl 3-Cyclohexyl-3-oxopropionate 95%

Lieferant: Apollo Scientific

OR965283-1G OR965283-5G
APOSOR965283-1GEA 0 EUR
APOSOR965283-1G APOSOR965283-5G
Ethyl 3-Cyclohexyl-3-oxopropionate 95%
Ethyl 3-Cyclohexyl-3-oxopropionate
Formel: C₁₁H₁₈O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04115574
CAS-Nummer: 15971-92-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR