Methyl-2-methyltetrahydrofuran-2-carboxylate 95%
Lieferant: Apollo Scientific
OR965309-1G
APOSOR965309-1GEA
487
EUR
APOSOR965309-1G
Methyl-2-methyltetrahydrofuran-2-carboxylate 95%
Methyl-2-methyltetrahydrofuran-2-carboxylate
Formel:
C₇H₁₂O₃ Molecular Weight: 144,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18651757 CAS-Nummer: 1218915-91-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...