3-Pyrrolidin-1-ylmethylbenzylamine 95%
Lieferant: Apollo Scientific
Synonyme:
(3-(Pyrrolidin-1-ylmethyl)phenyl)methanamine
OR965325-1G
APOSOR965325-1GEA
112
EUR
APOSOR965325-1G
3-Pyrrolidin-1-ylmethylbenzylamine 95%
3-Pyrrolidin-1-ylmethylbenzylamine
Formel:
C₁₂H₁₈N₂ Molecular Weight: 190,29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06408780 CAS-Nummer: 91271-78-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...