4-Oxo-3,4-dihydroquinazoline-2-carboxylic acid 95%
Lieferant: Apollo Scientific
Synonyme:
4-Quinazolone-2-carboxylic Acid, 4-Oxo-1,4-dihydroquinazoline-2-carboxylic acid
OR965326-1G
APOSOR965326-1GEA
278
EUR
APOSOR965326-1G
4-Oxo-3,4-dihydroquinazoline-2-carboxylic acid 95%
4-Oxo-3,4-dihydroquinazoline-2-carboxylic acid
Formel:
C₉H₆N₂O₃ Molecular Weight: 190,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02059339 CAS-Nummer: 29113-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...