2-(3-Nitro-phenyl)-1H-imidazole 95%

Lieferant: Apollo Scientific

OR965392-1G
APOSOR965392-1GEA 0 EUR
APOSOR965392-1G
2-(3-Nitro-phenyl)-1H-imidazole 95%
2-(3-Nitro-phenyl)-1H-imidazole
Formel: C₉H₇N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD05863370
CAS-Nummer: 13682-18-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR