3-Chloropyridazine-4-carbonitrile 95%

Lieferant: Apollo Scientific

OR965408-1G
APOSOR965408-1GEA 111 EUR
APOSOR965408-1G
3-Chloropyridazine-4-carbonitrile 95%
3-Chloropyridazine-4-carbonitrile
Formel: C5H2ClN3
Molecular Weight: 139,5400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11040160
CAS-Nummer: 1445-56-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR