4-(2-Aminoethyl)benzonitrile hydrochloride 97%

Lieferant: Apollo Scientific

OR965409-1G OR965409-5G
APOSOR965409-1GEA 145 EUR
APOSOR965409-1G APOSOR965409-5G
4-(2-Aminoethyl)benzonitrile hydrochloride 97%
4-(2-Aminoethyl)benzonitrile hydrochloride
Formel: C₉H₁₁ClN₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 167762-80-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR