(1R)-1-Pyridin-4-ylethanamine 95%

Lieferant: Apollo Scientific

Synonyme: (R)-1-(4-Pyridyl)ethylamine

OR965441-1G
APOSOR965441-1GEA 142 EUR
APOSOR965441-1G
(1R)-1-Pyridin-4-ylethanamine 95%
(1R)-1-Pyridin-4-ylethanamine
Formel: C₇H₁₀N₂
Molecular Weight: 122.168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09256818
CAS-Nummer: 45682-36-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR