(E)-2-Chloro-5-methylnicotinaldehyde oxime 96%

Lieferant: Apollo Scientific

OR965476-1G OR965476-5G
APOSOR965476-1GEA 219 EUR
APOSOR965476-1G APOSOR965476-5G
(E)-2-Chloro-5-methylnicotinaldehyde oxime 96%
(E)-2-Chloro-5-methylnicotinaldehyde oxime
Formel: C₇H₇ClN₂O
Molecular Weight: 170.6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13563057
CAS-Nummer: 1203500-13-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR