(E)-2-Chloro-5-methylnicotinaldehyde oxime 96%
Lieferant: Apollo Scientific
OR965476-1G
OR965476-5G
APOSOR965476-1GEA
219
EUR
APOSOR965476-1G
APOSOR965476-5G
(E)-2-Chloro-5-methylnicotinaldehyde oxime 96%
(E)-2-Chloro-5-methylnicotinaldehyde oxime
Formel:
C₇H₇ClN₂O Molecular Weight: 170.6 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13563057 CAS-Nummer: 1203500-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...