2-Pyrrolidin-1-ylbenzaldehyde 97%

Lieferant: Apollo Scientific
Gefahr

OR9655-1G OR9655-100MG OR9655-5G
APOSOR9655-1GEA 94.5 EUR
APOSOR9655-1G APOSOR9655-100MG APOSOR9655-5G
2-Pyrrolidin-1-ylbenzaldehyde 97%
2-Pyrrolidin-1-ylbenzaldehyde
Formel: C₁₁H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD03419322
CAS-Nummer: 58028-74-3
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR