4-(Cbz-amino)-1-Boc-piperidine-4-carboxylic acid 97%

Lieferant: Apollo Scientific

OR965510-1G OR965510-5G
APOSOR965510-1GEA 36.4 EUR
APOSOR965510-1G APOSOR965510-5G
4-(Cbz-amino)-1-Boc-piperidine-4-carboxylic acid 97%
4-(Cbz-amino)-1-Boc-piperidine-4-carboxylic acid
Formel: C₁₉H₂₆N₂O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD02683056
CAS-Nummer: 288154-16-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR