N-[4-(Dimethylamino)benzyl]-N-ethylamine 95%

Lieferant: Apollo Scientific

OR965584-1G OR965584-250MG OR965584-5G
APOSOR965584-1GEA 241 EUR
APOSOR965584-1G APOSOR965584-250MG APOSOR965584-5G
N-[4-(Dimethylamino)benzyl]-N-ethylamine 95%
N-[4-(Dimethylamino)benzyl]-N-ethylamine
Formel: C₁₁H₁₈N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00465457
CAS-Nummer: 313552-99-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR