4-Oxo-3,4-dihydro-phthalazine-1-carboxylic acid 95%
Lieferant: Apollo Scientific
Synonyme:
4-Oxo-3,4-dihydrophthalazine-1-carboxylic acid
OR965597-1G
APOSOR965597-1GEA
173
EUR
APOSOR965597-1G
4-Oxo-3,4-dihydro-phthalazine-1-carboxylic acid 95%
4-Oxo-3,4-dihydro-phthalazine-1-carboxylic acid
Formel:
C₉H₆N₂O₃ Molecular Weight: 190,156 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00524727 CAS-Nummer: 3260-44-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...