2-(4-Isobutylphenyl)acetic acid 95%

Lieferant: Apollo Scientific

Synonyme: Ibufenac

OR965605-1G
APOSOR965605-1GEA 63.8 EUR
APOSOR965605-1G
2-(4-Isobutylphenyl)acetic acid 95%
2-(4-Isobutylphenyl)acetic acid
Formel: C₁₂H₁₆O₂
Molecular Weight: 192.254 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00864374
CAS-Nummer: 1553-60-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR