3-(Cyanomethyl)benzoic acid 96%

Lieferant: Apollo Scientific

OR965652-1G OR965652-250MG
APOSOR965652-1GEA 45.1 EUR
APOSOR965652-1G APOSOR965652-250MG
3-(Cyanomethyl)benzoic acid 96%
3-(Cyanomethyl)benzoic acid
Formel: C9H7NO2
Lagertemperatur: Raumtemperatur
MDL: MFCD08234486
CAS-Nummer: 5689-33-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR