2-Chloro-1-(4-pyridinyl)ethanone hydrochloride 95%
Lieferant: Apollo Scientific
OR965660-1G
APOSOR965660-1GEA
487
EUR
APOSOR965660-1G
2-Chloro-1-(4-pyridinyl)ethanone hydrochloride 95%
2-Chloro-1-(4-pyridinyl)ethanone hydrochloride
Formel:
C₇H₆ClNO·ClH Molecular Weight: 192.043 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08692051 CAS-Nummer: 25260-36-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...