2-(Boc-amino)-4-methylpyridine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-[(tert-Butoxycarbonyl)amino]-4-methylpyridine, 2-(Boc-amino)-4-methylpyridine, (4-Methylpyridin-2-yl)carbamic acid tert-Butyl ester, tert-Butyl (4-Methylpyridin-2-yl)carbamate

OR965752-5G
APOSOR965752-5GEA 122 EUR
APOSOR965752-5G
2-(Boc-amino)-4-methylpyridine 95%
2-(Boc-amino)-4-methylpyridine
Formel: C₁₁H₁₆N₂O₂
Molecular Weight: 208.261 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07776933
CAS-Nummer: 90101-20-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR