4,4-Dimethyl-3,4-dihydro-2H-naphthalen-1-one 95%

Lieferant: Apollo Scientific

Synonyme: 4,4-Dimethyl-3,4-dihydronaphthalen-1(2H)-one

OR965754-1G
APOSOR965754-1GEA 545 EUR
APOSOR965754-1G
4,4-Dimethyl-3,4-dihydro-2H-naphthalen-1-one 95%
4,4-Dimethyl-3,4-dihydro-2H-naphthalen-1-one
Formel: C₁₂H₁₄O
Molecular Weight: 174,239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00089606
CAS-Nummer: 2979-69-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR