(±)-tert-Butyl-2-brombutyrat 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: tert-Butyl 2-bromobutanoate, tert-Butyl 2-bromobutyrate

OR965764-100G OR965764-25G
APOSOR965764-100GEA 481 EUR
APOSOR965764-100G APOSOR965764-25G
(±)-tert-Butyl-2-brombutyrat 98%
(±)-tert-Butyl-2-brombutyrat
Formel: C₈H₁₅BrO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00209634
CAS-Nummer: 24457-21-4
UN: 1760
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR