4,5-Dimethylisoxazole-3-carboxylic acid 95%

Lieferant: Apollo Scientific

OR965768-1G OR965768-250MG
APOSOR965768-1GEA 806 EUR
APOSOR965768-1G APOSOR965768-250MG
4,5-Dimethylisoxazole-3-carboxylic acid 95%
4,5-Dimethylisoxazole-3-carboxylic acid
Formel: C₆H₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD05667138
CAS-Nummer: 100047-61-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR