2-(2-Furyl)aniline 95%

Lieferant: Apollo Scientific
Achtung

OR9658-1G OR9658-250MG OR9658-500MG OR9658-5G
APOSOR9658-1GEA 0 EUR
APOSOR9658-1G APOSOR9658-250MG APOSOR9658-500MG APOSOR9658-5G
2-(2-Furyl)aniline 95%
2-(2-Furyl)aniline
Formel: C₁₀H₉NO
Lagertemperatur: Kühlschrank
MDL: MFCD04039070
CAS-Nummer: 55578-79-5
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR