8-Methoxy-2,3-dihydrobenzo[1,4]dioxine-6-carbaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 8-Methoxy-2,3-dihydrobenzo[b][1,4]dioxine-6-carbaldehyde

OR965808-1G
APOSOR965808-1GEA 116 EUR
APOSOR965808-1G
8-Methoxy-2,3-dihydrobenzo[1,4]dioxine-6-carbaldehyde 95%
8-Methoxy-2,3-dihydrobenzo[1,4]dioxine-6-carbaldehyde
Formel: C₁₀H₁₀O₄
Molecular Weight: 194,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06342801
CAS-Nummer: 75889-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR