2-(1-Methyl-1H-benzoimidazol-2-ylamino)-ethanol 96%
Lieferant: Apollo Scientific
Synonyme:
2-((1-Methyl-1H-benzo[d]imidazol-2-yl)amino)ethanol
OR965847-1G
APOSOR965847-1GEA
456
EUR
APOSOR965847-1G
2-(1-Methyl-1H-benzoimidazol-2-ylamino)-ethanol 96%
2-(1-Methyl-1H-benzoimidazol-2-ylamino)-ethanol
Formel:
C₁₀H₁₃N₃O Molecular Weight: 191.23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01136088 CAS-Nummer: 57262-39-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...