Ethyl-4-cyclopropyl-1,3-thiazole-2-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 4-cyclopropylthiazole-2-carboxylate

OR965854-1G
APOSOR965854-1GEA 301 EUR
APOSOR965854-1G
Ethyl-4-cyclopropyl-1,3-thiazole-2-carboxylate 95%
Ethyl-4-cyclopropyl-1,3-thiazole-2-carboxylate
Formel: C₉H₁₁NO₂S
Molecular Weight: 197.26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11656780
CAS-Nummer: 439692-05-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR