(2R)-2-(Propan-2-yl)pyrrolidine hydrochloride 97%

Lieferant: Apollo Scientific

Synonyme: (2R)-2-Propan-2-ylpyrrolidine, hydrochloride

OR965915-1G
APOSOR965915-1GEA 455 EUR
APOSOR965915-1G
(2R)-2-(Propan-2-yl)pyrrolidine hydrochloride 97%
(2R)-2-(Propan-2-yl)pyrrolidine hydrochloride
Formel: C₇H₁₅N·ClH
Molecular Weight: 149,662 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29037444
CAS-Nummer: 860640-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR