N-[2-(2-Furyl)benzyl]-N-methylamine

Lieferant: Apollo Scientific

OR9665-1G
APOSOR9665-1GEA 397 EUR
APOSOR9665-1G
N-[2-(2-Furyl)benzyl]-N-methylamine
N-[2-(2-Furyl)benzyl]-N-methylamine
Formel: C₁₂H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD08435880
CAS-Nummer: 859850-97-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR