4,5-Dibromo-2-chloropyridine ≥97%

Lieferant: Apollo Scientific

OR966917-250MG
APOSOR966917-250MGEA 72.5 EUR
APOSOR966917-250MG
4,5-Dibromo-2-chloropyridine ≥97%
4,5-Dibromo-2-chloropyridine
Formel: C₅H₂Br₂ClN
Molecular Weight: 271,337 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28750474
CAS-Nummer: 1807166-31-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR