4-(Methylsulphonyl)benzenesulphonyl chloride 95+%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(Methylsulphonyl)benzene-1-sulphonyl chloride

OR9678-250MG
APOSOR9678-250MGEA 21.9 EUR
APOSOR9678-250MG
4-(Methylsulphonyl)benzenesulphonyl chloride 95+%
4-(Methylsulphonyl)benzenesulphonyl chloride
Formel: C₇H₇ClO₄S₂
Molecular Weight: 254,699 g/mol
Schmelzpunkt: 165…169 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216486
CAS-Nummer: 82964-91-8
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR